Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ RECTAS
GREENER_CHOICESDP

Catalog No. 767950 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
76-795-0 50 mg
76-791-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 76-795-0 Supplier Tocris Bioscience™ Supplier No. 7679/50
Add to Cart
Add to Cart
This item is not returnable. View return policy

IKBKAP pre-mRNA splicing modulator

RECTAS (rectifier of aberrant splicing) is a pre-mRNA splicing modulator of the IKBKAP (inhibitor of κ light polypeptide gene enhancer in B-cells, kinase complex associated protein) gene. It corrects aberrant IKBKAP splicing in vitro in cellular models of familial dysautonomia and in vivo in transgenic mice. In combination with Phenylbutyric acid (Cat. No. 2682), RECTAS rescues PARK7 aberrant splicing and neuronal cell loss in a midbrain organoid model of familial Parkinson's disease. Orally bioavailable and brain penetrant.

Specifications

Chemical Name or Material 2-Chloro-N-(2-furanylmethyl)-7 H-purin-6-amine
CAS 101862-47-9
Quantity 50 mg
Target DNA, RNA and Protein Synthesis Compounds
Molecular Formula C10H8ClN5O
Purity 0.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.