Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RED Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | RED |
---|---|
Applications | Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
RED Polyclonal specifically detects RED in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
RED | |
Polyclonal | |
Rabbit | |
Cytokine Research | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
chondrosarcoma-associated protein 2, CSA2, Cytokine IK, IK cytokine, down-regulator of HLA II, IK factor, MGC59741, protein Red, Protein RER, RD element, REDprer protein, RER | |
IK | |
IgG | |
Affinity Purified |
Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
Human | |
Q13123 | |
3550 | |
This antibody was developed against a recombinant protein corresponding to amino acids: DMAVDSDEEVDYSKMDQGNKKGPLGRWDFDTQEEYSEYMNNKEALPKAAFQYGIKMSEGRKTRRFKETNDKAELDRQWKKISAIIEKRKKMEADGVEVKRPK | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title