Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Red Blood Cell Polyclonal Antibody, Rockland
SDP

Catalog No. 501057739
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-105-7739 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-105-7739 Supplier Rockland Immunochemicals Supplier No. 2094139
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Restore with 5.0 mL deionized water (or equivalent). Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use.

Red Blood Cell Polyclonal antibody specifically detects Red Blood Cell in Human samples. It is validated for Agglutination, Immunohistochemistry (Paraffin)

Specifications

Antigen Red Blood Cell
Applications Agglutination, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 10 mg/mL
Conjugate Unconjugated
Formulation 0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2
Host Species Rabbit
Immunogen Human washed pooled Red Blood Cells (RBC)
Purification Method Ion-exchange chromatography
Quantity 50 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.