Learn More
Red Blood Cell Polyclonal Antibody, Rockland

Rabbit Polyclonal Antibody
Supplier: Rockland Immunochemicals 2104139


Description
Restore with 5.0 mL deionized water (or equivalent). Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use.
Red Blood Cell Polyclonal antibody specifically detects Red Blood Cell in Mouse samples. It is validated for AgglutinationSpecifications
Red Blood Cell | |
Polyclonal | |
Unconjugated | |
Rabbit | |
Ion-exchange chromatography | |
RUO | |
Mouse | |
Lyophilized |
Agglutination | |
10 mg/mL | |
0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2 | |
Mouse washed pooled Red Blood Cells (RBC) | |
50 mg | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.