Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
3M™ Red Buffer Pad
Shop All 3M Products
Click to view available options
Diameter (Metric):
305 mm
356 mm
380 mm
406 mm
432 mm
460 mm
480 mm
530 mm
610 mm
Description
- Use to light scrub without dulling the finish
- Can be used for everyday cleaning under and automatic scrubber
- Also designed for spray buffing
- Cleans when damp; buffs when dry
- Use on rotary or automatic floor machines
- *Certified by Green Seal™ for Environmental Innovation based on a design for significantly improved initial performance compared to competitive products, extending the useful life of the floor pad and reducing solid waste generation
- *Certified by Green Seal™ for Environmental Innovation based on a design for significantly improved initial and mid-life performance compared to competitive products, extending the useful life of the floor pad and reducing solid waste generation

Specifications
Specifications
Product Type | Cleaning Pad |
Color | Red |
Diameter (Metric) | 480 mm |
For Use With (Application) | Buffing, Cleaning |
For Use With (Equipment) | Rotary or Automatic Floor Machines |
Material | Non-Woven Polyester Fiber |
Model | 5100 |
Quantity | 5/Cs. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction