Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Red Counterstain C
SDP

Catalog No. 48003019 Shop All Novus Products
Change view
Click to view available options
Quantity:
50 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
48-003-019 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 48-003-019 Supplier R&D Systems™ Supplier No. 48003019
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

R&D Systems™ Red Counterstain C is an acidophylic cytoplasmic counterstain that is included in the VasoTACS kit.

  • Provides an alternative red counterstain yielding a bright pink stain for vascular and smooth muscles tissues.
  • Hazard Statements (GHS-US):
    • H225 - Highly flammable liquid and vapor.
    • H319 - Causes serious eye irritation.
    • H371 - May cause damage to organs (optic nerve, central nervous system).
  • Precautionary Statements (GHS-US):
    • P210 - Keep away from extremely high or low temperatures, ignition sources, and incompatible materials. - No smoking
    • P233 - Keep container tightly closed.
    • P240 - Ground/bond container and receiving equipment
    • P241 - Use explosion-proof electrical, ventilating, and lighting equipment.
    • P242 - Use only non-sparking tools.
    • P243 - Take precautionary measures against static discharge.
    • P260 - Do not breathe vapors, mist, or spray.
    • P264 - Wash hands, forearms, and other exposed areas thoroughly after handling.
    • P270 - Do not eat, drink, or smoke when using this product.
    • P280 - Wear protective gloves, protective clothing, and eye protection.
    • P303+P361+P353 - IF ON SKIN (OR HAIR): Take off immediately all contaminated clothing. Rinse skin with water/shower.
    • P305+P351+P338 - IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    • P337+P313 - If eye irritation persists: Get medical advice/attention.
    • P370+P378 - In case of fire: Use appropriate media (see section 5) to extinguish.
    • P403+P235 - Store in a well-ventilated place. Keep cool.
    • P405 - Store locked up.
    • P501 - Dispose of contents/container in accordance with local, regional, national, territorial, provincial, and international regulations.

Specifications

Quantity 50 mL
Content And Storage Store the unopened product at room temperature
Product Line R&D Systems
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.