Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RED Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA550988
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA550988 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA550988 Supplier Invitrogen™ Supplier No. PA550988
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

IK Polyclonal Antibody detects endogenous levels of IK protein.

Insulin-like growth factor-II messenger RNA (mRNA)-binding protein-3 (IMP-3), also known as K homology domain-containing protein overexpressed in cancer (KOC) and L523S, is a member of the insulin-like growth factor-II mRNA-binding protein family and is expressed during embryogenesis and in some malignancies. IMP-3 is expressed in malignant melanoma but not in benign nevi, even when dysplastic features are present; IMP-3 is expressed in a significantly higher proportion of melanomas than Spitz nevi; and IMP-3 is expressed in metastatic melanomas significantly more than in thin melanomas. IMP-3 appears to be involved in the progression of malignant melanoma and may play an important role in the regulation of the biologic behavior of this tumor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RED
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4
Gene IK
Gene Accession No. Q13123, Q66HG8, Q9Z1M8
Gene Alias arginine/glutamic acid/aspartic acid repeat containing protein; chondrosarcoma-associated protein 2; CSA2; Cytokine IK; Cytokine IK factor; IK; IK cytokine; IK cytokine, down-regulator of HLA II; IK factor; MuRED; prer protein; protein Red; Protein RER; RD element; RED; Rer
Gene Symbols IK
Host Species Rabbit
Immunogen Synthesized peptide derived from the C-terminal region of human IK.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 24010, 291659, 3550
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.