Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RED Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA532151
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA532151 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA532151 Supplier Invitrogen™ Supplier No. PA532151
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: 293T, A431, HepG2. Predicted reactivity: Mouse (96%), Rat (96%), Chicken (87%), Rhesus Monkey (98%), Chimpanzee (99%), Bovine (97%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Insulin-like growth factor-II messenger RNA (mRNA)-binding protein-3 (IMP-3), also known as K homology domain-containing protein overexpressed in cancer (KOC) and L523S, is a member of the insulin-like growth factor-II mRNA-binding protein family and is expressed during embryogenesis and in some malignancies. IMP-3 is expressed in malignant melanoma but not in benign nevi, even when dysplastic features are present; IMP-3 is expressed in a significantly higher proportion of melanomas than Spitz nevi; and IMP-3 is expressed in metastatic melanomas significantly more than in thin melanomas. IMP-3 appears to be involved in the progression of malignant melanoma and may play an important role in the regulation of the biologic behavior of this tumor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RED
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.01 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.01% thimerosal; pH 7
Gene IK
Gene Accession No. Q13123
Gene Alias arginine/glutamic acid/aspartic acid repeat containing protein; chondrosarcoma-associated protein 2; CSA2; Cytokine IK; Cytokine IK factor; IK; IK cytokine; IK cytokine, down-regulator of HLA II; IK factor; MuRED; prer protein; protein Red; Protein RER; RD element; RED; Rer
Gene Symbols IK
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human RED. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3550
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.