Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Plasticoid Red Rubber Sleeve Stoppers for Serum and Vaccine Bottles
Shop All Plasticoid Products

Click to view available options
Size:
11.5
13
14.5
15
Stopper Number:
11.5
13
14.5
19
Length (Metric) Sleeve:
1cm
1.46cm
1.5cm
1.9cm
Description
- Comprised of dry natural rubber
- Folds down over neck of bottle
- With hollow plugs, rounded corners and needle guide
- Easily perforated with hypodermic needles
- Self-sealing
Specifications
Specifications
Product Type | Sleeve Stopper |
Stopper Number | 13 |
Size | 13 |
Length (Metric) Sleeve | 1.46cm |
Diameter (Metric) Plug | 1cm |
Material | Rubber |
For Use With (Equipment) | Serum and vaccine bottles |
Color | Red |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction