Learn More
Description
- Color of this sample is due to oxidized iron becoming interspersed with sand grains during lithification
- For use in comparing white sandstone to understand physical expression of chemical differences
- Correlates well to Next Generation Science Standards Evidence Statements
- Red color due to inclusion of iron in chemical structure of rock
Specifications
Specifications
| Product Type | Sedimentary Rock, Red Sandstone |
| For Use With (Application) | Comparing white sandstone to understand physical expression of chemical differences |
| Packaging | Clear labeled and barcoded bag |
| Quantity | 1 Ea. |
| Specimen Size | 2 to 3 cm |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
