Learn More
Eisco™ Red Sandstone Specimen
SCI_ED
Catalog No. S26592 Shop All Eisco Products
Change view
Click to view available options
Quantity:
1 Ea.
10/Pk.
12/Pk.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S26592 10/Pk.
S24730 1 Ea.
S24733 12/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. S26592 Supplier Eisco™ Supplier No. ESNG0035PK10
Add to Cart
Add to Cart
Red Sandstone Specimen is made up of well-sorted and well-rounded grains of quartz.
  • Color of this sample is due to oxidized iron becoming interspersed with sand grains during lithification
  • For use in comparing white sandstone to understand physical expression of chemical differences
  • Correlates well to Next Generation Science Standards Evidence Statements
  • Red color due to inclusion of iron in chemical structure of rock
Specifications
Product Type Sedimentary Rock
Quantity 10/Pk.
Specimen Size 2 to 3 cm
Weight (Metric) 0.2 kg
Specimen Type Red Sandstone