Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Safetec™ Red-Z™ Biohazard Fluid Control Powders
SDP

Catalog No. p-37125
Click to view available options
Type:
Green Z Spill Control Solidifier
Red Z Spill Control Solidifier
Packaging Quantity:
5 oz.
11 oz.
15 oz.
3.5 lb.

For safe cleanup of spills of bodily fluids

Total fluid control encapsulators solidify and deodorize blood, vomit, urine or other potentially hazardous, water-based fluids. 

  • Red-Z solidifies and contains chlorine to deodorize potentially infectious blood and bodily fluids
  • Green-Z solidifies and can be used in the cleanup of blood, bodily fluids, chemotherapy, glutaraldehyde, and mercury spills

 

Order Info

Supplied in shaker-top bottles.

Specifications

Certifications/Compliance OSHA
Type Red Z Spill Control Solidifier
Format Powder
For Use With (Application) Solidify and deodorize potentially Infectious blood and body fluids
Packaging Quantity 15 oz.
Weight (Metric) 0.42 kg
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.