Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

REDD1 Mouse anti-Human, Clone: 1G11, Bethyl Laboratories

Catalog No. 501571216
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-157-1216 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-157-1216 Supplier Bethyl Laboratories, Inc Supplier No. A500001AT
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

RTP801 was initially identified as a gene induced by DNA damage, and later found to also be regulated by other cellular stresses such as hypoxia and glucocorticoid treatment. Recently, RTP801 has been shown to act as a mediator of tuberous sclerosis complex (TSC)-dependent regulation of the mammalian Target of Rapamycin (mTOR), an evolutionarily conserved serine/threonine kinase that regulates cell growth and cell cycle. In response to energy stress, RTP801 inhibits mTOR function, resulting in dephosphorylation of downstream targets such as ribosomal protein S6 kinase 1 and 4EBP1 and decreasing cell growth. Disregulation of RTP801 may thus contribute to human tumorigenesis.

Specifications

Antigen REDD1
Applications Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 1G11
Conjugate Unconjugated
Formulation phosphate, tris citrate with 0.09% sodium azide; pH 7-8
Gene DDIT4
Gene Accession No. Q9NX09
Gene Alias Protein regulated in development and DNA damage response 1; REDD-1; HIF-1 responsive protein RTP801
Gene Symbols DDIT4
Host Species Mouse
Immunogen Human C-Reaction Protein
Purification Method Affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54541
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.