Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Reduced Folate Carrier/SLC19A1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP159904
Description
Reduced Folate Carrier/SLC19A1 Polyclonal specifically detects Reduced Folate Carrier/SLC19A1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Reduced Folate Carrier/SLC19A1 | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
CHMD, FLOT1, folate transporter 1, FOLThuman reduced folate carrier (RFC)10Intestinal folate carrier 1, IFC1, IFC-1, Placental folate transporter, Reduced folate carrier protein, REFC, RFC, RFC1, solute carrier family 19 (folate transporter), member 1, Solute carrier family 19 member 1 | |
Rabbit | |
Protein A purified | |
RUO | |
6573 | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 100μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
1 mg/ml | |
Western Blot 1.0 ug/ml, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500 | |
P41440 | |
SLC19A1 | |
Synthetic peptides corresponding to SLC19A1(solute carrier family 19 (folate transporter), member 1) The peptide sequence was selected from the N terminal of SLC19A1. Peptide sequence MVPSSPAVEKQVPVEPGPDPELRSWRHLVCYLCFYGFMAQIRPGESFITP. | |
100 μL | |
Primary | |
Expected identity based on immunogen sequence: Human: 100%. | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction