Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Reelin Antibody (CL5324), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP261414
Description
Reelin Monoclonal specifically detects Reelin in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Reelin | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
RELN | |
This antibody was developed against a recombinant protein corresponding to amino acids: TFCEPYGPRELITTGLNTTTASVLQFSIGSGSCRFSYSDPSIIVLYAKNNSADWIQLEKIRAPSNVSTIIHILYLPEDAKGENVQFQWKQENLRVGEVYE | |
100 μL | |
5649 | |
Human, Mouse, Rat | |
Purified |
Immunohistochemistry (Paraffin) | |
CL5324 | |
Immunohistochemistry-Paraffin 1:50 - 1:200 | |
EC 3.4.21, EC 3.4.21.-, reelin, RLPRO1598 | |
Mouse | |
Protein A purified | |
Primary | |
Specificity of human Reelin antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction