Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Reelin Antibody (RELN/4498), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP314203F
This item is not returnable.
View return policy
Description
Reelin Monoclonal antibody specifically detects Reelin in Human, Mouse, Mammalia samples. It is validated for ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
Reelin | |
Monoclonal | |
FITC | |
PBS | |
Mouse | |
Protein A or G purified | |
RUO | |
5649 | |
Store at 4°C in the dark. | |
Purified |
ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
RELN/4498 | |
ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
EC 3.4.21, EC 3.4.21.-, reelin, RLPRO1598 | |
Recombinant fragment (around aa 1796-2115) of human Reelin protein (Uniprot: P78509) | |
0.1 mL | |
Primary | |
Human, Mouse, Mammalia | |
Antibody | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction