Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Reelin C-Term Mouse anti-Mouse, Clone: 12C10, MilliporeSigma™

Catalog No. MABN2428100 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABN2428100 100 μL
MABN242825 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABN2428100 Supplier MilliporeSigma Supplier No. MABN2428100UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reelin C-Term Monoclonal antibody specifically detects Reelin C-Term in Mouse samples. It is validated for Immunofluorescence, Immunohistochemistry, Inhibition, Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Reelin C-Term
Applications Immunofluorescence, Immunohistochemistry, Inhibition Assays, Western Blot
Classification Monoclonal
Clone 12C10
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation Purified mouse monoclonal antibody IgG2b in PBS without azide.
Host Species Mouse
Immunogen KLH-conjugated linear peptide corresponding to 17 amino acids from the C-terminal region of human Reelin.
Purification Method Protein G purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Stable for 1 year at -10°C to -25°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.