Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ REEP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100315
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100315 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100315 Supplier Invitrogen™ Supplier No. PA5100315
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total REEP1.

REEP1 belongs to the DP1 family and may enhance the cell surface expression of odorant receptors.
TRUSTED_SUSTAINABILITY

Specifications

Antigen REEP1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Reep1
Gene Accession No. Q8BGH4, Q9H902
Gene Alias C2orf23; D6Ertd253e; HMN5B; rcG_56072; receptor accessory protein 1; receptor expression-enhancing protein 1; receptor expression-enhancing protein 1-like; Reep1; RGD1305230; si:dkey-21a6.4; Spastic paraplegia 31 protein; SPG31; Yip2a
Gene Symbols Reep1
Host Species Rabbit
Immunogen A synthesized peptide derived from human REEP1(Accession Q9H902), corresponding to amino acid residues R160-A201.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 362384, 52250, 65055
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.