Learn More
Invitrogen™ REEP1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5121145
Description
Positive test controls include: Mouse brain, Mouse spinal cord, Rat brain. The target is usually found in the following locations: Endoplasmic reticulum, Membrane, Mitochondrion membrane, Multi-pass membrane protein. Immunogen sequence: KEIDDCLVQA KDRSYDALVH FGKRGLNVAA TAAVMAASKG QGALSERLRS FSMQDLTTIR GDGAPAPSGP PPPGSGRASG KHGQPKMSRS ASESASSSGT A.
REEP1 belongs to the DP1 family and may enhance the cell surface expression of odorant receptors.
Specifications
REEP1 | |
Polyclonal | |
Unconjugated | |
Reep1 | |
C2orf23; D6Ertd253e; HMN5B; rcG_56072; receptor accessory protein 1; receptor expression-enhancing protein 1; receptor expression-enhancing protein 1-like; Reep1; RGD1305230; si:dkey-21a6.4; Spastic paraplegia 31 protein; SPG31; Yip2a | |
Rabbit | |
Affinity Chromatography | |
RUO | |
362384, 52250, 65055 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
2.36 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8BGH4, Q9H902 | |
Reep1 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 101-201 of human REEP1 (NP_075063.1). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.