Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
REEP2 Antibody (S326D-29), Novus Biologicals™

Mouse Monoclonal Antibody
$548.00
Specifications
Antigen | REEP2 |
---|---|
Clone | S326D-29 |
Concentration | 0.5 mg/ml |
Dilution | Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:500 |
Classification | Monoclonal |
Description
REEP2 Monoclonal specifically detects REEP2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
REEP2 | |
0.5 mg/ml | |
Monoclonal | |
Purified | |
RUO | |
C5orf19receptor expression-enhancing protein 2, chromosome 5 open reading frame 19, receptor accessory protein 2, SGC32445receptor expression enhancing protein 2 | |
REEP2 | |
IgG1 | |
Protein G purified |
S326D-29 | |
Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:500 | |
Unconjugated | |
Mouse | |
Human, Mouse, Rat | |
51308 | |
Fusion protein amino acids 111-254 (Cytoplasmic C-terminus) of mouse REEP2. | |
Primary | |
Detects 30kDa. Cross-reacts with REEP1. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title