Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ REEP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116724
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116724 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116724 Supplier Invitrogen™ Supplier No. PA5116724
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: RD, Mouse brain, Mouse testis, Rat testis Immunogen sequence: HPTLSNKEKE IDEYITQARD KSYETMMRVG KRGLNLAANA AVTAAAKGVL SEKLRSFSMQ DLTLIRDEDA LPLQRPDGRL RPSPGSLLDT IEDLGDDPAL SLRSSTNPAD SRTEASEDDM GDKAPKRAKP IKKAPKAEPL ASKTLKTRPK KKTSGGGDSA.

Mammalian odorant receptors require accessory proteins such as RTP1 and RTP2 for functional cell surface expression. Receptor expression-enhancing protein (REEP) family members are transmembrane proteins which interact with odorant receptor proteins and may enhance the odorant receptor responses to odorants. Recently studies have shown other chemosensory receptors such as bitter taste receptors are also influenced by RTP and REEP family members.
TRUSTED_SUSTAINABILITY

Specifications

Antigen REEP2
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.53 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene REEP2
Gene Accession No. Q8VCD6
Gene Alias C2orf19; C5orf19; LOC532432 protein; LOC682105; receptor accessory protein 2; receptor expression-enhancing protein 2; Reep2; SGC32445; SPG72; Yip2d
Gene Symbols REEP2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 93-252 of human REEP2 (NP_0576902).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 225362, 682105
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.