Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
REEP4 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | REEP4 |
---|---|
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
REEP4 Polyclonal specifically detects REEP4 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
REEP4 | |
Polyclonal | |
Rabbit | |
Human | |
Q9H6H4 | |
80346 | |
This antibody was developed against a recombinant protein corresponding to amino acids: LRSISDAPAPAYHDPLYLEDQVSHRRPPIGYRAGGLQDSDTEDECWSDTEAVPRAPARPREKPLIRSQSLRVVKRKPPVREGTSRSLKVRTRKKTVPSDVD | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence | |
Unconjugated | |
RUO | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
C8orf20chromosome 8 open reading frame 20, FLJ22246, FLJ22277, PP432, receptor accessory protein 4, receptor expression enhancing protein 4, receptor expression-enhancing protein 4 | |
REEP4 | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title