Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
REEP4 Antibody, PE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP230361PE
Description
REEP4 Polyclonal antibody specifically detects REEP4 in Human samples. It is validated for Immunoprecipitation, Western Blot (Negative)Specifications
REEP4 | |
Polyclonal | |
Immunoprecipitation, Western Blot (Negative) | |
C8orf20chromosome 8 open reading frame 20, FLJ22246, FLJ22277, PP432, receptor accessory protein 4, receptor expression enhancing protein 4, receptor expression-enhancing protein 4 | |
Maps to a region between residue 200 to 225 of human Receptor Accessory Protein 4 using the numbering given in entry NP_079508.2 (GeneID 80346). | |
0.1 mL | |
Signal Transduction | |
80346 | |
Store at 4C in the dark. | |
IgG |
Immunoprecipitation | |
PE | |
PBS | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction