Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ REEP4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA520885
Description
A suggested positive control is human lung tissue lysate. PA5-20885 can be used with blocking peptide PEP-0999.
Microtubule-binding protein required to ensure proper cell division and nuclear envelope reassembly by sequestering the endoplasmic reticulum away from chromosomes during mitosis. Probably acts by clearing the endoplasmic reticulum membrane from metaphase chromosomes. [UniProt].
Specifications
REEP4 | |
Polyclonal | |
Unconjugated | |
REEP4 | |
2700029E10Rik; C8orf20; PP432; receptor accessory protein 4; receptor expression enhancing protein 4; receptor expression-enhancing protein 4; Reep4; RGD1306561; Yip2c | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
306014, 72549, 80346 | |
Maintain refrigerated at 2°C to 8°C for up to 3 months. For long term storage store at -20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
Q4QQW1, Q8K072, Q9H6H4 | |
REEP4 | |
A 17 amino acid peptide near the center of human REEP4. | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction