Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ REEP4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560079
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560079 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560079 Supplier Invitrogen™ Supplier No. PA560079
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LRSISDAPAP AYHDPLYLED QVSHRRPPIG YRAGGLQDSD TEDECWSDTE AVPRAPARPR EKPLIRSQSL RVVKRKPPVR EGTSRSLKVR TRKKTVPSDV D Highest antigen sequence identity to the following orthologs: Mouse - 80%, Rat - 81%.

Microtubule-binding protein required to ensure proper cell division and nuclear envelope reassembly by sequestering the endoplasmic reticulum away from chromosomes during mitosis. Probably acts by clearing the endoplasmic reticulum membrane from metaphase chromosomes. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen REEP4
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene REEP4
Gene Accession No. Q9H6H4
Gene Alias 2700029E10Rik; C8orf20; PP432; receptor accessory protein 4; receptor expression enhancing protein 4; receptor expression-enhancing protein 4; Reep4; RGD1306561; Yip2c
Gene Symbols REEP4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human REEP4. Recombinant protein control fragment (Product #RP-99895).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80346
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.