Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
REEP5 Antibody (OTI4D2), PE/Cy7, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273850PECY7
Description
REEP5 Monoclonal specifically detects REEP5 in Human, Mouse, Rat samples. It is validated for Flow Cytometry.Specifications
REEP5 | |
Monoclonal | |
PE-Cyanine7 | |
PBS with 0.05% Sodium Azide | |
REEP5 | |
Full length recombinant protein of human REEP5 (NP_005660) produced in HEK293T cells. | |
0.1 mL | |
7905.0 | |
Store at 4°C in the dark. Do not freeze. |
Flow Cytometry | |
OTI4D2 | |
Flow Cytometry | |
C5orf18polyposis coli region hypothetical protein DP1, chromosome 5 open reading frame 18, D5S346deleted in polyposis 1, DP1TB2YOP1, MGC70440, Polyposis locus protein 1, Protein TB2, receptor accessory protein 5, receptor expression enhancing protein 5, receptor expression-enhancing protein 5 | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction