Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

REEP5 Antibody (OTI4D2), PerCP, Novus Biologicals™
SDP

Catalog No. NB273850PCP Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB273850PCP 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB273850PCP Supplier Novus Biologicals Supplier No. NBP273850PCP
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

REEP5 Monoclonal specifically detects REEP5 in Human, Mouse, Rat samples. It is validated for Flow Cytometry.

Specifications

Antigen REEP5
Applications Flow Cytometry
Classification Monoclonal
Clone OTI4D2
Conjugate PerCP
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias C5orf18polyposis coli region hypothetical protein DP1, chromosome 5 open reading frame 18, D5S346deleted in polyposis 1, DP1TB2YOP1, MGC70440, Polyposis locus protein 1, Protein TB2, receptor accessory protein 5, receptor expression enhancing protein 5, receptor expression-enhancing protein 5
Gene Symbols REEP5
Host Species Mouse
Immunogen Full length recombinant protein of human REEP5 (NP_005660) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 7905.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.