Learn More
Invitrogen™ REEP5 Monoclonal Antibody (OTI4D2)

Description
REEP5 Monoclonal Antibody for Western Blot, IHC (P), Flow
Specifications
Specifications
| Antigen | REEP5 |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | OTI4D2 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | REEP5 |
| Gene Accession No. | Q00765 |
| Gene Alias | AI324241; AU022809; AW495741; C5orf18; D5S346; deleted in polyposis 1; Dp1; DP1/TB2; GP106; polyposis coli region hypothetical protein DP1; polyposis locus protein 1; Polyposis locus protein 1 homolog; Protein TB2; protein TB2 homolog; R74856; receptor accessory protein 5; receptor expression enhancing protein 5; receptor expression-enhancing protein 5; Reep5; TB2; TB2/DP1; Yip2e; YOP1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.