Learn More
REEP5 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$151.31
Specifications
Antigen | REEP5 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Liquid |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-157-0639
![]() |
Bethyl Laboratories, Inc
A305517AT |
20 μL |
Each of 1 for $151.31
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Bovine and Orangutan
May promote functional cell surface expression of olfactory receptors.Specifications
REEP5 | |
Polyclonal | |
Liquid | |
RUO | |
TBS with 0.1% BSA and 0.09% sodium azide | |
REEP5, C5orf18, D5S346, Deleted in polyposis 1, Deleted in polyposis, Polyposis locus protein 1, Protein TB2, Receptor accessory protein 5, TB2, YOP1, DP1 | |
Between 139 and 189 | |
Primary | |
4° C | |
REEP5 |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
Q00765, Q60870 | |
13476, 7905 | |
IgG | |
Antigen affinity chromatography | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.