Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ REEP6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA546611
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA546611 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA546611 Supplier Invitrogen™ Supplier No. PA546611
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: LYQRVVRPLF LRHHGAVDRI MNDLSGRALD AAAGITRNVK PSQTPQPKDK Sequence homology: Human: 100%.

REEP6 may enhance the cell surface expression of odorant receptors.
TRUSTED_SUSTAINABILITY

Specifications

Antigen REEP6
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene REEP6
Gene Accession No. Q96HR9
Gene Alias 0610011M24Rik; C19orf32; deleted in polyposis 1-like 1; DP1L1; Polyposis locus protein 1-like 1; receptor accessory protein 6; receptor expression enhancing protein 6; receptor expression-enhancing protein 6; REEP6; TB2 protein-like 1; Tb2l1; Yip2f
Gene Symbols REEP6
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human REEP6.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 92840
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.