Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rees and Ecker Blood Diluting Fluid, for Platelet Count, Ricca Chemical

Click to view available options
Quantity:
120 mL
500 mL
Packaging:
Natural Poly Bottle
Specifications
Specifications
Physical Form | Liquid |
Chemical Name or Material | Rees and Ecker Blood Diluting Fluid |
Name Note | For Platelet Count |
CAS | 7732-18-5, 6132-04-3, 50-00-0, 81029-05-2, 67-56-1 |
CAS Min % | 94.0, 3.58, 0.2, 0.05 |
CAS Max % | 98.0, 3.73, 0.21, 0.05 |
Packaging | Natural Poly Bottle |
Color | Blue |
Quantity | 120 mL |
Grade | Laboratory |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction