Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Reflectoquant™ Ammonium Test Strips
Click to view available options
Graduations:
0.2 to 7mg/L
5 to 20mg/L
Description
For accurate and precise on-site analysis.
- Available using the indophenol blue or the Nessler method
- Use in agriculture, food or water analysis, hygiene monitoring, industrial analysis
Order Info
50 tests per kit

Specifications
Specifications
For Use With (Application) | The test measures both ammonium ions and dissolved ammonia in e.g. groundwater and surface water, seawater, drinking water and aquarium water |
Graduations | 0.2 to 7mg/L |
Quantity | 50 Tests |
Storage Requirements | 2°C to 8°C |
Test Method | Indophenol blue |
Type | Ammonium Test Strip |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction