Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Refractory Metals, plasma standard solution, Specpure™, Al, B, Cr, Hf, Mo, Nb, Si, Ta, Ti, V, W, Zr at 100µg/mL

Manufacturer:  Alfa Aesar™ 44270AE

Quantity 100mL

 View more versions of this product

Catalog No. AA44270AE


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

MFCD00151265
UN3264
Refractory Metals plasma standard solution, Al, B, Cr, Hf, Mo, Nb, Si, Ta, Ti, V, W, Zr at 100μg/mL
Specpure™

Specifications

Liquid
UN3264
Matrix: 5% HCl/tr. HF
Specpure™
MFCD00151265
100mL
Refractory Metals plasma standard solution, Al, B, Cr, Hf, Mo, Nb, Si, Ta, Ti, V, W, Zr at 100μg/mL

Safety and Handling

SDS & Safety Documents

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, INORGANIC, N.O.S.

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit