Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Reg1A Antibody (431202), Alexa Fluor™ 350, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals FAB4937U
Description
Reg1A Monoclonal antibody specifically detects Reg1A in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOFSpecifications
Reg1A | |
Monoclonal | |
Alexa Fluor 350 | |
50mM Sodium Borate | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Flow Cytometry, CyTOF | |
431202 | |
Western Blot, Intracellular Staining by Flow Cytometry, CyTOF-ready | |
ICRF, Islet cells regeneration factor, Islet of Langerhans regenerating protein, lithostathine-1-alpha, P19, pancreatic stone protein, secretory, Pancreatic thread protein, protein-X, PSP, PSPS, PSPS1, PTP, REG, REG-1-alpha, Regenerating islet-derived protein 1-alpha, Regenerating protein I alpha | |
E. coli-derived recombinant human Reg1A, Gln23-Asn166, Accession # P05451 | |
0.1 mL | |
Cell Cycle and Replication | |
5967 | |
Store at 4C in the dark. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction