Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ REG1A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561038
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561038 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561038 Supplier Invitrogen™ Supplier No. PA561038
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TDDFNVWIGL HDPKKNRRWH WSSGSLVSYK SWG Highest antigen sequence identity to the following orthologs: Mouse - 79%, Rat - 76%.

This gene is a type I subclass member of the Reg gene family. The Reg gene family is a multigene family grouped into four subclasses, types I, II, III and IV, based on the primary structures of the encoded proteins. This gene encodes a protein that is secreted by the exocrine pancreas. It is associated with islet cell regeneration and diabetogenesis and may be involved in pancreatic lithogenesis. Reg family members REG1B, REGL, PAP and this gene are tandemly clustered on chromosome 2p12 and may have arisen from the same ancestral gene by gene duplication.
TRUSTED_SUSTAINABILITY

Specifications

Antigen REG1A
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene REG1A
Gene Accession No. P05451
Gene Alias ICRF; islet cells regeneration factor; Islet of Langerhans regenerating protein; LITHOST; Lithostathine; lithostathine 1 alpha; lithostathine-1-alpha; MGC12447; P19; pancreatic stone protein; pancreatic stone protein, secretory; pancreatic thread protein; protein-X; PSP; PSPS; PSPS1; PTP; RATLITHOST; RATRGPI; Reg; Reg1; REG1A; REG-1-alpha; regenerating family member 1 alpha; regenerating islet-derived 1 alpha; regenerating islet-derived mouse homolog 1; regenerating islet-derived protein 1-alpha; Regenerating protein I alpha; regeneration protein, lithostatin, pancreatic stone protein; Rgp1; RGPI
Gene Symbols REG1A
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human REG1A. Recombinant protein control fragment (Product #RP-99484).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5967
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.