Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
REG1A Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
$191.80 - $427.00
Specifications
Antigen | REG1A |
---|---|
Concentration | 0.5 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
FEPA5145307
|
Invitrogen™
PA5145307 |
200 μL |
Each for $427.00
|
|
|||||
FEPA5145308
|
Invitrogen™
PA5145308 |
20 μL |
Each for $191.80
|
|
|||||
Description
This gene is a type I subclass member of the Reg gene family. The Reg gene family is a multigene family grouped into four subclasses, types I, II, III and IV, based on the primary structures of the encoded proteins. This gene encodes a protein that is secreted by the exocrine pancreas. It is associated with islet cell regeneration and diabetogenesis and may be involved in pancreatic lithogenesis. Reg family members REG1B, REGL, PAP and this gene are tandemly clustered on chromosome 2p12 and may have arisen from the same ancestral gene by gene duplication.Specifications
REG1A | |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
Mouse, Rat | |
P10758 | |
24714 | |
Recombinant Rat REG1a protein, Gln22-Ala165 (Accession #P10758). | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
0.5 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4 | |
ICRF; islet cells regeneration factor; Islet of Langerhans regenerating protein; LITHOST; Lithostathine; lithostathine 1 alpha; lithostathine-1-alpha; MGC12447; P19; pancreatic stone protein; pancreatic stone protein, secretory; pancreatic thread protein; protein-X; PSP; PSPS; PSPS1; PTP; RATLITHOST; RATRGPI; Reg; Reg1; REG1A; REG-1-alpha; regenerating family member 1 alpha; regenerating islet-derived 1 alpha; regenerating islet-derived mouse homolog 1; regenerating islet-derived protein 1-alpha; Regenerating protein I alpha; regeneration protein, lithostatin, pancreatic stone protein; Rgp1; RGPI | |
REG1A | |
IgG | |
Protein A | |
REG1A |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title