Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Reg1B Antibody (008), mFluor Violet 500 SE, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289937MFV500
Description
Reg1B Monoclonal antibody specifically detects Reg1B in Human samples. It is validated for ELISA, ELISASpecifications
Reg1B | |
Monoclonal | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
5968 | |
Store at 4C in the dark. | |
IgG |
ELISA | |
008 | |
ELISA, Sandwich ELISA Capture | |
lithostathine-1-beta, Pancreatic stone protein 2, pancreatic threadprotein), PSPS2PSP-2, regenerating islet-derived 1 beta, Regenerating islet-derived protein 1-beta, Regenerating protein I beta, REGH, REGLREG-1-beta, secretory pancreatic stone protein 2 | |
Rabbit immunized with purified, recombinant Human Reg1B (Accession#: NP_006498.1, Met1-Asn166) | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction