Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Reg1B Antibody (103), DyLight 405, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289940V
Description
Reg1B Monoclonal specifically detects Reg1B in Human samples. It is validated for ELISA, Sandwich ELISA Detection.Specifications
Reg1B | |
Monoclonal | |
DyLight 405 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
5968 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
103 | |
ELISA, Sandwich ELISA Detection | |
lithostathine-1-beta, Pancreatic stone protein 2, pancreatic threadprotein), PSPS2PSP-2, regenerating islet-derived 1 beta, Regenerating islet-derived protein 1-beta, Regenerating protein I beta, REGH, REGLREG-1-beta, secretory pancreatic stone protein 2 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human Reg1B (NP_006498.1; Met 1-Asn 166). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction