Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ REG2 Polyclonal Antibody
Goat Polyclonal Antibody
Supplier: Invitrogen™ PA547242
Description
In Western blots, approximately 50% cross-reactivity with recombinant mouse Reg1 is observed and less than 1% cross-reactivity with recombinant human (rh) Reg1B, recombinant rat (rr) Reg2, rrReg3 and rhReg4 is observed. Reconstitute at 0.2 mg/mL in sterile PBS.

Specifications
REG2 | |
Polyclonal | |
Unconjugated | |
Reg2 | |
islet of Langerhans regenerating protein 2; lithostathine 1 beta; lithostathine-1-beta; Lithostathine-2; pancreatic stone protein 2; Pancreatic thread protein 2; PSP; PSP-2; PSPS2; PTP; REG1B; REG-1-beta; Reg2; REG-2; regenerating family member 1 beta; regenerating islet-derived 1 beta (pancreatic stone protein, pancreatic thread protein); regenerating islet-derived 2; regenerating islet-derived protein 1-beta; regenerating islet-derived, mouse homolog 2; regenerating protein I beta; REGH; REGI-BETA; RegII; REGL; secretory pancreatic stone protein 2 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
19693 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry, Western Blot | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
Q08731 | |
Reg2 | |
E. coli-derived recombinant mouse Reg2 Asp35-Ala173. | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction