Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Reg3A Antibody (010), Alexa Fluor™ 488, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290468AF488
Description
Reg3A Monoclonal specifically detects Reg3A in Mouse samples. It is validated for ELISA.Specifications
Reg3A | |
Monoclonal | |
Alexa Fluor 488 | |
hepatocarcinoma-intestine-pancreas, HIPpancreatitis-associated protein, Human proislet peptide, pancreatic beta cell growth factor, Pancreatitis-associated protein 1, PAP homologous protein, PAP1FLJ18565, PAPPAP-H, PBCGF, proliferation-inducing protein 34, proliferation-inducing protein 42, Reg III-alpha, REG3, REG-3-alpha, regenerating islet-derived 3 alpha, regenerating islet-derived protein 3-alpha, Regenerating islet-derived protein III-alpha, REG-III | |
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Reg3A (NP_035389.1; Met1-Gln175). | |
0.1 mL | |
Primary | |
Mouse | |
Purified |
ELISA | |
10 | |
50 mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
5068 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction