Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

REG3A Mouse anti-Human, Clone: 8B11.1, MilliporeSigma™

Catalog No. MABF824100 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF824-100 100 μL
MABF824-25UL 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABF824-100 Supplier MilliporeSigma Supplier No. MABF824100UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-REG3A, clone 8B11.1 is a mouse monoclonal antibody that detects Regenerating islet-derived protein 3-alpha (REG3A) and has been tested for use in Immunohistochemistry (Paraffin) and Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen REG3A
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 8B11.1
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Gene Symbols REG3A,PAP,PAP1
Host Species Mouse
Immunogen GST/His-tagged recombinant fragment corresponding to 128 amino acids from the internal region of human Regenerating islet-derived protein 3-alpha.
Purification Method Protein G Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Inflammation and Immunology
Primary or Secondary Primary
Gene ID (Entrez) NP_002571
Test Specificity Clone 8B11.1 is a mouse monoclonal antibody that detects human Regenerating islet-derived protein 3-alpha.
Target Species Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.