Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ REG3B Monoclonal Antibody (293523)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA523999
Description
This antibody does not cross-react with recombinant mouse Reg1A, 1B, 2A, recombinant rat Reg3, or recombinant human Reg4. Reconstitute at 0.5 mg/mL in sterile PBS.

Specifications
REG3B | |
Monoclonal | |
0.5 mg/mL | |
PBS with 5% trehalose and No Preservative | |
P25031 | |
Reg3b | |
E. coli-derived recombinant rat Reg3B Glu27-Gly175. | |
100 μg | |
Primary | |
Rat | |
Antibody | |
IgG1 |
Immunohistochemistry (Frozen), Western Blot | |
293523 | |
Unconjugated | |
Reg3b | |
HIP; Pancreatitis-associated protein 1; Pap; PAP1; peptide 23; Reg III-beta; Reg2; REG-2; Reg3b; REG-3-beta; regenerating islet-derived 3 beta; regenerating islet-derived protein 3 beta; regenerating islet-derived protein 3-beta; Regenerating islet-derived protein 3-beta 15 kDa form; Regenerating islet-derived protein 3-beta 16.5 kDa form; Regenerating islet-derived protein III-beta; REG-III; RegIII (beta) | |
Mouse | |
Protein A/G | |
RUO | |
24618 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction