Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ REG3B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA547700
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA547700 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA547700 Supplier Invitrogen™ Supplier No. PA547700
Add to Cart
Add to Cart

Sheep Polyclonal Antibody

In direct ELISAs, approximately 75% cross-reactivity with recombinant rat (rr) Reg3B is observed. In Western blots, approximately 30% cross-reactivity with rrReg3B is observed. In direct ELISAs and Western blots, less than 1% cross-reactivity with rrReg3A, recombinant mouse (rm) Reg3A, and rmReg3G is observed. Reconstitute at 0.2 mg/mL in sterile PBS.

The REG3B gene encodes a protein that is part of the regenerating islet-derived (Reg) family, with significant roles in antimicrobial activity and tissue repair. REG3B is primarily expressed in the pancreas and is implicated in pancreatic regeneration, playing a critical role in conditions such as diabetes and pancreatitis. It exhibits potential therapeutic benefits in tissue injury and inflammatory disorders, including inflammatory bowel disease and acute liver injury. REG3B promotes acinar-to-ductal metaplasia through the activation of the RAS-RAF-MEK-ERK signaling pathway, suggesting its involvement in the early stages of pancreatic ductal adenocarcinoma development. Additionally, REG3B has been identified as an essential regulator of macrophage trafficking in cardiac healing post-myocardial ischemia, aiding in the recruitment and expansion of myeloid cells. Research aims to target REG3B and its receptors to interrupt pathological processes, offering promising avenues for treatment of pancreatic and extrapancreatic diseases.
TRUSTED_SUSTAINABILITY

Specifications

Antigen REG3B
Applications Immunohistochemistry (Frozen), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene Reg3b
Gene Accession No. P35230
Gene Alias HIP; Pancreatitis-associated protein 1; Pap; PAP1; peptide 23; Reg III-beta; Reg2; REG-2; Reg3b; REG-3-beta; regenerating islet-derived 3 beta; regenerating islet-derived protein 3 beta; regenerating islet-derived protein 3-beta; Regenerating islet-derived protein 3-beta 15 kDa form; Regenerating islet-derived protein 3-beta 16.5 kDa form; Regenerating islet-derived protein III-beta; REG-III; RegIII (beta)
Gene Symbols Reg3b
Host Species Sheep
Immunogen E. coli-derived recombinant mouse Reg3B Glu27-Gly175.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18489
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.