Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Reg3G Antibody (002), HRP , Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289944H
Description
Reg3G Monoclonal antibody specifically detects Reg3G in Human samples. It is validated for ELISASpecifications
Reg3G | |
Monoclonal | |
HRP | |
LPPM429, MGC118999, MGC119001, Pancreatitis-associated protein 1B, Pancreatitis-associated protein IB, PAP IB, PAP-1B, PAP1BMGC118998, PAPIB, Reg III-gamma, REG-3-gamma, regenerating gene III, regenerating islet-derived 3 gamma, regenerating islet-derived protein 3-gamma, Regenerating islet-derived protein III-gamma, REG-III, UNQ429 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human Reg3G (NP_001008388.1; Met 1-Asp 175). | |
0.1 mL | |
Primary | |
Human | |
Purified |
ELISA | |
002 | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
130120 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction