Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Reg4 Antibody (018), Biotin , Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289831B
Description
Reg4 Monoclonal antibody specifically detects Reg4 in Human samples. It is validated for ELISASpecifications
Reg4 | |
Monoclonal | |
Biotin | |
Gastrointestinal secretory protein, GISPREG-4, Reg IV, regenerating gene type IV, regenerating islet-derived family, member 4, regenerating islet-derived protein 4, Regenerating islet-derived protein IV, REG-IV, RELPREG-like protein | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human Reg4 (NP_001152824.1; Met 1-Pro 158). | |
0.1 mL | |
Immunology | |
83998 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
018 | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction