Learn More
Description
Specifications
Specifications
| Cell Line | The lysate was created in HEK293T cells, using plasmid ID RC2228 and based on accession number NM_3244. The protein contains a C-terminal DDK tag. |
| For Use With (Application) | Western Blot |
| Storage Buffer | RIPA buffer |
| Purity | Protein |
| Test Specificity | Homo sapiens regenerating islet-derived family, member 4 (REG4), mRNA. |
| Content And Storage | -80°C. Avoid freeze-thaw cycles. |
| Research Category | Immunology |
| Protein State | Native |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
