Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Regucalcin Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody has been used in 1 publication

$382.00 - $610.00

Specifications

Antigen Regucalcin
Dilution Western Blot 0.4 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 1:50-1:200
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB434096
SDP
View Documents
Novus Biologicals
NBP18084925UL
25ul
Each for $382.00
Only null left
Add to Cart
 
NBP180849
SDP
View Documents
Novus Biologicals
NBP180849
0.1 mL
Each for $610.00
Only null left
Add to Cart
 
Description

Description

Regucalcin Polyclonal specifically detects Regucalcin in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

Regucalcin
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
RUO
EC 3.1.1.17, Gluconolactonase, GNL, RCsenescence marker protein-30, regucalcin (senescence marker protein-30), Senescence marker protein 30, SMP-30, SMP30regucalcin
RGN
IgG
Affinity Purified
Specificity of human Regucalcin antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Western Blot 0.4 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 1:50-1:200
Polyclonal
Rabbit
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
9104
This antibody was developed against Recombinant Protein corresponding to amino acids:DAEGKLWVACYNGGRVIRLDPVTGKRLQTVKLPVDKTTSCCFGGKNYSEMYVTCARDGMDPEGLLRQPEAGGIFKITGLGVKGIAPYSY
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.