Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Corning™ Regular Fetal Bovine Serum

Click to view available options
Quantity:
500 mL
Product Type:
Fetal Bovine Serum
Heat Inactivated:
No
Yes
pH:
7.6 ±0.8
7.8 ±0.8
Description
- Sera is aseptically collected and processed in a cGMP facility
- Each batch of raw serum is screened for endotoxin, residual hemoglobin concentration, and microbiological quality before being used in production
- Batches of raw serum are pooled and continuously mixed prior to filtration to ensure consistent quality throughout the entire lot
- Final lots are tested for endotoxin, hemoglobin, pH, mycoplasma, and growth promotion. Biochemical assays and virus testing are also available upon request
- Regular FBS is sourced from herds in USDA approved countries (ex: Mexico, Honduras, Costa Rica, Nicaragua, etc.)
Order Info
Shipped in frozen form

Specifications
Specifications
Certifications/Compliance | cGMP |
Color | Amber Yellow to Reddish |
Content And Storage | -25° to -15°C |
Product Type | Fetal Bovine Serum |
Quantity | 500 mL |
Sterility | Sterile |
Sterilization Method | Triple-filtered, 0.1 μm |
Source | Bovine Fetus |
Grade | Regular |
Endotoxin Concentration | ≤20EU/ mL |
Show More |
For research use only.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction