Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ Regular Grade Top Grain Cowhide Leather Drivers Gloves

Catalog No. 19121019 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
X-Small
Medium
Large
X-Large
2X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-121-019 X-Small
19-121-019B Medium
19-121-019C Large
19-121-019D X-Large
19-121-019E 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 19-121-019 Supplier PIP™ Supplier No. 68163/XS
Add to Cart
Add to Cart
This item is not returnable. View return policy

Top grain cowhide leather construction provides comfort, durability, abrasion resistance, and breathability. Drivers gloves feature a slip-on cuff to enhance fit and comfort.

  • Top grain cowhide leather is smooth, provides durability and dexterity
  • Keystone thumb is ergonomic, reduces hand fatigue, offers dexterity and durability
  • Elastic closure sewn inside glove for safety and comfort
  • Cotton color binding denotes sizing
  • Meets the content requirements for Chromium VI free leather

Specifications

Color Natural
Grade Regular
Hand Specific
Material Top Grain Cowhide Leather
Product Type Driver Gloves
Size X-Small
Style Standard
For Use With (Application) Construction, Machine Operation, Light Duty Welding, Utility Work, Farming, and Roping
Glove Cuff Style Open
Hem Color Light Blue
Series 68-163
Thumb Style Keystone
Length (English) 8.7 in.
Length (Metric) 22 cm
Weight 26.5 lb. (Case)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.