Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Plas Labs™ Regular Stream Table Complete with Land Form System
SDPSCI_ED

Catalog No. S68804 Shop All Plas-Labs, Inc. Products
Click to view available options
:
Plas Lab™ Stream Table Set
This item is not returnable. View return policy
This item is not returnable. View return policy

Stream Table kit that is designed and created to facilitate the simulations and study of many basic land forms for Earth Science.

Kit that is designed and created to facilitate the simulations and study of many basic land forms for Earth Science.

  • Includes the table, wave maker, rainfall simulator, and countour grid.
  • 60' long x 20' wide x 6' deep

Specifications

Grade 6+
For Use With (Application) Facilitates the simulations and study of many basic land forms for Earth Science with extra accessories
Product Type Teaching Model
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.